3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%, Thermo Scientific Chemicals
3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H55144.MD
also known as H55144-MD
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
73.60 
Save 10.95 (15%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
73.60 
Save 10.95 (15%)
Add to cart
3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%, Thermo Scientific Chemicals
Catalog numberH55144.MD
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
73.60 
Save 10.95 (15%)
-
Add to cart
Specifications
Chemical Name or Material3,3'-Diethyl-9-methylthiacarbocyanine iodide
CAS3065-79-0
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only