Thermo Scientific Chemicals

6-Fluoronicotinic acid, 97%, Thermo Scientific Chemicals

Catalog number: H55147.03
1 g, Each
Thermo Scientific Chemicals

6-Fluoronicotinic acid, 97%, Thermo Scientific Chemicals

Catalog number: H55147.03
1 g, Each
Quantity
Catalog number: H55147.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
403-45-2
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥96.0% (GC)
GC/MS Analysis
Conforms

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text