2,4-Dichloro-5-methylpyrimidine, is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine. It can react with piperidineto produce 2-chloro-5-methyl-4-piperidin-1-yl-pyrimidine. This reaction will need solvent dioxane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dichloro-5-methylpyrimidine, is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine. It can react with piperidineto produce 2-chloro-5-methyl-4-piperidin-1-yl-pyrimidine. This reaction will need solvent dioxane.
Solubility
Soluble in chloroform, ether, ethyl acetate and toluene
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Henry C. Koppel.; Robert Henre Springer.; Roland K. Robins.; C. C. Cheng. Pyrimidines. V. Analogs of 2-(o-Chlorobenzylthio)-4-dimethylamino- 5-methylpyrimidine (Bayer DG-428)1. J. Org. Chem. 1962, 27, 91), 181-185.
- Bryan W. Roberts.; Joseph B. Lambert.; John D. Roberts. Nitrogen-15 Magnetic Resonance Spectroscopy. VI. Pyrimidine Derivatives1,2. J. Am. Chem. Soc. 1965, 87, (23), 5439-5441.