Thermo Scientific Chemicals

2-(Boc-amino)ethyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: H55211.03
1 g, Each
Thermo Scientific Chemicals

2-(Boc-amino)ethyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: H55211.03
1 g, Each
Quantity
Catalog number: H55211.03
also known as H55211-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39684-80-5
IUPAC Name
tert-butyl N-(2-bromoethyl)carbamate
Molecular Formula
C7H14BrNO2
InChI Key
TZRQZPMQUXEZMC-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCCBr
Appearance (Color)
White to pale yellow
Form
Low melting solid or Powder
Assay (GC)
?95%
Proton NMR
Confirms

Description

N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store at -20°C. Reactive with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text