It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.
Solubility
Soluble in Chloroform, Dichloromethane, Ethanol and Ethyl Ether.
Notes
Air sensitive. Keep under 2-8°C. Store under inert gas.
RUO – Research Use Only
Laurent Gomez.; Michael D. Hack.; Jiejun Wu.; John J.M. Wiener.; Hari Venkatesan.; Alejandro Santillán Jr.; Daniel J. Pippel.; Neelakandha Mani.; Brian J. Morrow.; S. Timothy Motley.; Karen Joy Shaw.; Ronald Wolin.; Cheryl A. Grice.; Todd K. Jones. Novel pyrazole derivatives as potent inhibitors of type II topoisomerases. Part 1: Synthesis and preliminary SAR analysis.Bioorg. Med. Chem. Lett. 2007, 17 (10), 2723-2727.