Thermo Scientific Chemicals

Methylaminoacetonitrile hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: H55215.06
5 g, Each
Thermo Scientific Chemicals

Methylaminoacetonitrile hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: H55215.06
5 g, Each
Quantity
Catalog number: H55215.06
also known as H55215-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25808-30-4
IUPAC Name
hydrogen 2-(methylamino)acetonitrile chloride
Molecular Formula
C3H7ClN2
InChI Key
DFKBQHBEROHUNF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CNCC#N
Assay (unspecified)
>98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text