Thermo Scientific Chemicals

2-Amino-5-bromobenzonitrile, 96%, Thermo Scientific Chemicals

Catalog number: H55217.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-5-bromobenzonitrile, 96%, Thermo Scientific Chemicals

Catalog number: H55217.06
5 g, Each
Quantity
Catalog number: H55217.06
also known as H55217-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39263-32-6
IUPAC Name
2-amino-5-bromobenzonitrile
Molecular Formula
C7H5BrN2
InChI Key
OATYCBHROMXWJO-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=C1C#N
Assay (HPLC)
>95.0%

Description

2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text