N-Boc-aniline, 97%, Thermo Scientific Chemicals
N-Boc-aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55232.06
also known as H55232-06
Price (USD)/ Each
40.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
40.40
Add to cart
N-Boc-aniline, 97%, Thermo Scientific Chemicals
Catalog numberH55232.06
Price (USD)/ Each
40.40
-
Add to cart
Chemical Identifiers
CAS3422-01-3
IUPAC Nametert-butyl N-phenylcarbamate
Molecular FormulaC11H15NO2
InChI KeyKZZHPWMVEVZEFG-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormFused solid
Assay (GC)≥96.0%
Proton NMRConforms
Carbon-13 NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only