It is a useful aromatic compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful aromatic compound.
Solubility
Slightly soluble in water.
Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
RUO – Research Use Only
General References:
- Naoyuki Shimada.; Taiki Hanari.; Yasunobu Kurosaki.; Masahiro Anada.; Hisanori Nambu.; Shunichi Hashimoto. Catalytic asymmetric synthesis of descurainin via 1,3-dipolar cycloaddition of a carbonyl ylide using Rh2(R-TCPTTL)4.Tetrahedron Lett. 2010, 51 (50), 6572-6575.
- Masahiko Yamaguchi.; Yukiharu Tsukamoto.; Chinatsu Ikeura.; Shigeo Nakamura.; Toru Minami. Regioselective Synthesis and Polymerization of Substituted o-Trimethylsilylphenylacetylenes. Chemistry Letters. 1991, 20 (7),Chemistry Letters.