2-(Tri-n-butylstannyl)furan, 97%, Thermo Scientific Chemicals
2-(Tri-n-butylstannyl)furan, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Tri-n-butylstannyl)furan, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H55259.AA
also known as H55259-AA
Price (USD)/ Each
31.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
37.30 
Save 5.65 (15%)
-
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)/ Each
31.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
37.30 
Save 5.65 (15%)
Add to cart
2-(Tri-n-butylstannyl)furan, 97%, Thermo Scientific Chemicals
Catalog numberH55259.AA
Price (USD)/ Each
31.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
37.30 
Save 5.65 (15%)
-
Add to cart
Chemical Identifiers
CAS118486-94-5
IUPAC Nametributyl(furan-2-yl)stannane
Molecular FormulaC16H30OSn
InChI KeySANWDQJIWZEKOD-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Assay (GC)>96.0%
It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Store at room temperature. Light sensitive.
RUO – Research Use Only

General References:

  1. Yun Yang.; Henry N.C. Wong. Regiospecific synthesis of 3,4-disubstituted furans and 3-substituted furans using 3,4-Bis(tri-n-butylstannyl)furan and 3-(tri-n-butylstannyl)f. Tetrahedron. 1994, 50 (32),9583-9608.
  2. Donald MacLeod.; David Moorcroft.; Peter Quayle Michael R.J. Dorrity.; John F. Malone.; Gareth M. Davies. The Pd(0)-catalysed coupling reactions of 2-(tri-n-butylstannyl)-3,4-dihydrofuran and -5,6-dihydropyran. Tetrahedron Lett. 1990, 31 (42),6077-6080.