Thermo Scientific Chemicals

2-Amino-5-bromobenzothiazole, 97%, Thermo Scientific Chemicals

Catalog number: H55270.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-5-bromobenzothiazole, 97%, Thermo Scientific Chemicals

Catalog number: H55270.03
1 g, Each
Quantity
Catalog number: H55270.03
also known as H55270-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20358-03-6
IUPAC Name
5-bromo-1,3-benzothiazol-2-amine
Molecular Formula
C7H5BrN2S
InChI Key
ZPUJTWBWSOOMRP-UHFFFAOYSA-N
SMILES
NC1=NC2=CC(Br)=CC=C2S1
Assay (HPLC)
>96.0%

Description

2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text