Thermo Scientific Chemicals

1-Bromo-3,5-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: H55312.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-3,5-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: H55312.14
25 g, Each
Quantity
Catalog number: H55312.14
also known as H55312-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20469-65-2
Assay (HPLC)
>96.0%

Description

1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.

Solubility
Soluble in methanol.

Notes
Reacts with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text