Thermo Scientific Chemicals

4-Nitrobenzenediazonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: H55315.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrobenzenediazonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: H55315.14
25 g, Each
Quantity
Catalog number: H55315.14
also known as H55315-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
456-27-9
IUPAC Name
4-nitrobenzene-1-diazonium; tetrafluoroboranuide
Molecular Formula
C6H4BF4N3O2
InChI Key
AFULQCYCQAHYIP-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N
Assay (HPLC)
>96.0%

Description

4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4'-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

Solubility
Soluble in acetonitrile and dimethyl sulfoxide.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text