Thermo Scientific Chemicals

Di(2-pyridyl) ketone, 98%, Thermo Scientific Chemicals

Catalog number: H55322.14
25 g, Each
Thermo Scientific Chemicals

Di(2-pyridyl) ketone, 98%, Thermo Scientific Chemicals

Catalog number: H55322.14
25 g, Each
Quantity
Catalog number: H55322.14
also known as H55322-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19437-26-4
IUPAC Name
2-(pyridine-2-carbonyl)pyridine
Molecular Formula
C11H8N2O
InChI Key
QPOWUYJWCJRLEE-UHFFFAOYSA-N
SMILES
O=C(C1=CC=CC=N1)C1=CC=CC=N1
Appearance (Color)
White to cream to pale brown
Assay (chromatographic, GC or HPLC)
≥97.5%
Form
Powder or crystalline powder

Description

di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(?-OR)(6)}(3+) complexes with a star topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(μ-OR)(6)}(3+) complexes with a “star” topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

Solubility
Low solubility in water.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text