1-(4-Chlorobenzyl)piperazine, 98%, Thermo Scientific Chemicals
1-(4-Chlorobenzyl)piperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Chlorobenzyl)piperazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H55337.03
also known as H55337-03
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.10 
Save 4.45 (14%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.10 
Save 4.45 (14%)
Add to cart
1-(4-Chlorobenzyl)piperazine, 98%, Thermo Scientific Chemicals
Catalog numberH55337.03
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.10 
Save 4.45 (14%)
-
Add to cart
Chemical Identifiers
CAS23145-88-2
IUPAC Name1-[(4-chlorophenyl)methyl]piperazine
Molecular FormulaC11H15ClN2
InChI KeyGSJXJZOWHSTWOX-UHFFFAOYSA-N
SMILESClC1=CC=C(CN2CCNCC2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only