Thermo Scientific Chemicals

trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl, 97%, Thermo Scientific Chemicals

Catalog number: H55356.03
1 g, Each
Thermo Scientific Chemicals

trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl, 97%, Thermo Scientific Chemicals

Catalog number: H55356.03
1 g, Each
Quantity
Catalog number: H55356.03
also known as H55356-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82832-57-3
IUPAC Name
4-(3,4-difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
Molecular Formula
C21H30F2
InChI Key
FSWZOZXLWVWJAH-UHFFFAOYSA-N
SMILES
CCCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C=C1
Assay (GC)
>96.0%

Description

It is  used in liquid crystal monomer, pharmaceutical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is  used in liquid crystal monomer, pharmaceutical synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text