Thermo Scientific Chemicals

2-Amino-7-bromofluorene, tech. 90%, Thermo Scientific Chemicals

Catalog number: H55370.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-7-bromofluorene, tech. 90%, Thermo Scientific Chemicals

Catalog number: H55370.06
5 g, Each
Quantity
Catalog number: H55370.06
also known as H55370-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6638-60-4
IUPAC Name
7-bromo-9H-fluoren-2-amine
Molecular Formula
C13H10BrN
InChI Key
RJWYTISHBMNMOZ-UHFFFAOYSA-N
SMILES
NC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1
Assay (HPLC)
>88.0%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Sparingly soluble in water. (0.724 mg/L)

Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text