9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H55373.03
also known as H55373-03
Price (USD)/ Each
125.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
125.00
Add to cart
9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH55373.03
Price (USD)/ Each
125.00
-
Add to cart
Chemical Identifiers
CAS258865-48-4
IUPAC Name[7-(dihydroxyboranyl)-9,9-dioctyl-9H-fluoren-2-yl]boronic acid
Molecular FormulaC29H44B2O4
InChI KeyHURJMQMZDPOUOU-UHFFFAOYSA-N
SMILESCCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMR>96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only