Thermo Scientific Chemicals

Pyrrole-3-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: H55383.MD
250 mg, Each
Thermo Scientific Chemicals

Pyrrole-3-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: H55383.MD
250 mg, Each
Quantity
Catalog number: H55383.MD
also known as H55383-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
931-03-3
IUPAC Name
1H-pyrrole-3-carboxylic acid
Molecular Formula
C5H5NO2
InChI Key
DOYOPBSXEIZLRE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CNC=C1
Form
Off-white to yellow
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥98.0%

Description

Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

Solubility
Soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text