5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
Solubility
Soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Dan Peters.; Anna-Britta Hornfeldt.; Salo Gronowitz. Synthesis of various 5-substituted uracils. Journal of Heterocyclic Chemistry. 1990, 27, (7), 2165-2173.