Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals
Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55395.03
also known as H55395-03
Price (USD)/ Each
49.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
49.70
Add to cart
Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals
Catalog numberH55395.03
Price (USD)/ Each
49.70
-
Add to cart
Chemical Identifiers
CAS26305-75-9
IUPAC Nameλ¹-cobalt(1+) tris(triphenylphosphane) chloride
Molecular FormulaC54H45ClCoP3
InChI KeyRHNBOEDTKLKUEB-UHFFFAOYSA-M
SMILES[Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.0% min
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive.
RUO – Research Use Only

General References:

  1. Den-Ichi Momose.; Kazuo Iguchi.; Toshikazu Sugiyama.; Yasuji Yamada. Reductive coupling of allylic halides by chlorotris(triphenylphosphine)cobalt(I).Tetrahedron Lett. 1983, 24 (9),921-924.
  2. Den-ichi Momose.; Yasuji Yamada. Reaction of bromohydrins with chlorotris(triphenylphosphine)cobalt (I). Tetrahedron Lett. 1983, 24 (26),2669-2672.