Thermo Scientific Chemicals

Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals

Catalog number: H55395.06
5 g, Each
Thermo Scientific Chemicals

Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals

Catalog number: H55395.06
5 g, Each
Quantity
Catalog number: H55395.06
also known as H55395-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26305-75-9
IUPAC Name
λ¹-cobalt(1+) tris(triphenylphosphane) chloride
Molecular Formula
C54H45ClCoP3
InChI Key
RHNBOEDTKLKUEB-UHFFFAOYSA-M
SMILES
[Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
96.0% min

Description

A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text