Thermo Scientific Chemicals

Bis(2-aminophenyl) ether, 98%, Thermo Scientific Chemicals

Catalog number: H55396.06
5 g, Each
Thermo Scientific Chemicals

Bis(2-aminophenyl) ether, 98%, Thermo Scientific Chemicals

Catalog number: H55396.06
5 g, Each
Quantity
Catalog number: H55396.06
also known as H55396-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24878-25-9
IUPAC Name
2-(2-aminophenoxy)aniline
Molecular Formula
C12H12N2O
InChI Key
GOJFAKBEASOYNM-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1OC1=CC=CC=C1N
Proton NMR
Conforms
Appearance (Color)
White to brown
Form
Powder
Assay (HPLC)
≥97.5%

Description

It is catalytic reagent and used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is catalytic reagent and used in pharmaceutical industry.

Solubility
Slightly soluble in water.

Notes
Store at low temperature, away from light and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text