Thermo Scientific Chemicals

3-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H55436.03
1 g, Each
Thermo Scientific Chemicals

3-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H55436.03
1 g, Each
Quantity
Catalog number: H55436.03
also known as H55436-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76006-33-2
IUPAC Name
3-bromo-2-methylbenzoic acid
Molecular Formula
C8H7BrO2
InChI Key
BJGKVCKGUBYULR-UHFFFAOYSA-N
SMILES
CC1=C(C=CC=C1Br)C(O)=O
Assay (HPLC)
>96.0%

Description

3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

Solubility
Soluble in chloroform, methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text