3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.
Solubility
Soluble in chloroform, methanol
Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
RUO – Research Use Only
W. S. Lindsay.; Peter Stokes.; Leslie G. Humber.; V. Boekelheide. Syntheses of 4,12-Dimethyl[2.2]metacyclophane. J. Am. Chem. Soc. 1961, 83 (4),943-949.
Xiangshu Xiao.; Smitha Antony.; Yves Pommier.; Mark Cushman. Total Synthesis and Biological Evaluation of 22-Hydroxyacuminatine.J. Med. Chem. 2006, 49 (4),1408-1412.