Chemical Identifiers
CAS4727-50-8
IUPAC Name1-ethyl-4-[3-(1-ethyl-1,4-dihydroquinolin-4-ylidene)prop-1-en-1-yl]quinolin-1-ium iodide
Molecular FormulaC25H25IN2
InChI KeyCEJANLKHJMMNQB-UHFFFAOYSA-M
SMILES[I-].CCN1C=CC(=CC=CC2=C3C=CC=CC3=[N+](CC)C=C2)C2=CC=CC=C12
View more