Thermo Scientific Chemicals

3-n-Octylthiophene, 97%, Thermo Scientific Chemicals

Catalog number: H55466.14
25 g, Each
Thermo Scientific Chemicals

3-n-Octylthiophene, 97%, Thermo Scientific Chemicals

Catalog number: H55466.14
25 g, Each
Quantity
Catalog number: H55466.14
also known as H55466-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65016-62-8
IUPAC Name
3-octylthiophene
Molecular Formula
C12H20S
InChI Key
WQYWXQCOYRZFAV-UHFFFAOYSA-N
SMILES
CCCCCCCCC1=CSC=C1
Assay (HPLC)
>96.0%

Description

3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text