N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.
Notes
Light Sensitive. Avoid oxidizing agents.
RUO – Research Use Only
General References:
- Herbert C. Brown.; Marek Zaidlewicz.; Pramod V. Dalvi. Molecular Addition Compounds. 10. Borane Adducts with N,N-Dialkylanilines for Hydroboration. Organometallics. 1998, 17 (19),4202-4205.
- Nadiya G Korzhenevska.; Volodymyr I Rybachenko.; Grzegorz Schroeder. The basicity of 1,8-bis(dimethylamino)naphthalene and the hybrid state of the nitrogen atoms of its dimethylamino groups. Tetrahedron Lett. 2002, 43 (34),6043-6045.