4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Zeng, Y.; Moghadam, P. Z.; Snurr, R. Q. Pore Size Dependence of Adsorption and Separation of Thiophene/Benzene Mixtures in Zeolites. J. Phys. Chem. C 2015, 119 (27), 15263-15273.
Zhang, L.; Fu, W.; Xiang, M.; Wang, W.; He, M.; Tang, T. MgO Nanosheet Assemblies Supported CoMo Catalyst with High Activity in Hydrodesulfurization of Dibenzothiophene. Ind. Eng. Chem. Res. 2015, 54 (21), 5580-5588.