Thermo Scientific Chemicals

Tricyclohexyltin chloride, 97%, Thermo Scientific Chemicals

Catalog number: H55493.03
1 g, Each
Thermo Scientific Chemicals

Tricyclohexyltin chloride, 97%, Thermo Scientific Chemicals

Catalog number: H55493.03
1 g, Each
Quantity
Catalog number: H55493.03
also known as H55493-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3091-32-5
Molecular Formula
C18H35ClSn
InChI Key
WLSJDIYSPLULMS-UHFFFAOYSA-N
SMILES
Cl.C1CCC(CC1)[SnH](C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol)
405.64
Form
Solid
Purity
≥96.0%

Description

Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text