Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Sirajuddin, M.; Ali, S.; Haider, A.; Shah, N. A.; Shah, A.; Khan, M. R. Synthesis, characterization, biological screenings and interaction with calf thymus DNA as well as electrochemical studies of adducts formed by azomethine [2-((3, 5-dimethylphenylimino)methyl)phenol] and organotin(IV) chlorides. Polyhedron 2012, 40 (1), 19-31.
Toupance, T.; Renard, L.; Jousseaume, B.; Olivier, C.; Pinoie, V.; Verbruggen, I.; Willem, R. Silica-anchored organotin trichloride: a recyclable and clean organotin catalyst for transesterification reactions. Dalton Trans. 2013, 42 (26), 9764-9770.