Thermo Scientific Chemicals

4-(2-Pyridyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H55498.06
5 g, Each
Thermo Scientific Chemicals

4-(2-Pyridyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H55498.06
5 g, Each
Quantity
Catalog number: H55498.06
also known as H55498-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127406-56-8
IUPAC Name
4-(pyridin-2-yl)benzaldehyde
Molecular Formula
C12H9NO
InChI Key
NMLYGLCBSFKJFI-UHFFFAOYSA-N
SMILES
O=CC1=CC=C(C=C1)C1=CC=CC=N1
Assay (HPLC)
>97.5%

Description

4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir

Solubility
Soluble in Chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text