2-Amino-4,6-difluorobenzothiazole, 97%, Thermo Scientific Chemicals
2-Amino-4,6-difluorobenzothiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4,6-difluorobenzothiazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H55506.06
also known as H55506-06
Price (USD)/ Each
88.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
88.80
Add to cart
2-Amino-4,6-difluorobenzothiazole, 97%, Thermo Scientific Chemicals
Catalog numberH55506.06
Price (USD)/ Each
88.80
-
Add to cart
Specifications
Chemical Name or Material2-Amino-4,6-difluorobenzothiazole
CAS119256-40-5
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-Amino-4,6-difluorobenzothiazole is used as drugs, food, cosmetics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4,6-difluorobenzothiazole is used as drugs, food, cosmetics

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only