Diphenylborinic anhydride, 95%, Thermo Scientific Chemicals
Diphenylborinic anhydride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylborinic anhydride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H55509.MD
also known as H55509-MD
Price (USD)/ Each
69.40
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
69.40
Add to cart
Diphenylborinic anhydride, 95%, Thermo Scientific Chemicals
Catalog numberH55509.MD
Price (USD)/ Each
69.40
-
Add to cart
Chemical Identifiers
CAS4426-21-5
IUPAC Namediphenylboranyl diphenylborinate
Molecular FormulaC24H20B2O
InChI KeySNQFEECGHGUHBK-UHFFFAOYSA-N
SMILESO(B(C1=CC=CC=C1)C1=CC=CC=C1)B(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystal
Assay by NMR≥ 94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only