3,3-Dimethylbutyraldehyde, 97%, Thermo Scientific Chemicals
3,3-Dimethylbutyraldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethylbutyraldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 mL
Catalog number H55522.AA
also known as H55522-AA
Price (USD)/ Each
217.00
-
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)/ Each
217.00
Add to cart
3,3-Dimethylbutyraldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH55522.AA
Price (USD)/ Each
217.00
-
Add to cart
Chemical Identifiers
CAS2987-16-8
IUPAC Name3,3-dimethylbutanal
Molecular FormulaC6H12O
InChI KeyLTNUSYNQZJZUSY-UHFFFAOYSA-N
SMILESCC(C)(C)CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only