(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals
(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55534.03
also known as H55534-03
Price (USD)/ Each
32.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
32.00
Add to cart
(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals
Catalog numberH55534.03
Price (USD)/ Each
32.00
-
Add to cart
Chemical Identifiers
CAS16116-78-2
IUPAC Name[2-(4-bromophenyl)ethynyl]trimethylsilane
Molecular FormulaC11H13BrSi
InChI KeyRNMSGCJGNJYDNS-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#CC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
It is used as the intermediate of Organic Light Emitting Diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as the intermediate of Organic Light Emitting Diode (OLED).

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Birgit Wieczorek.; Annika Träff.; Patrik Krumlinde.; Harm P. Dijkstra.; Maarten R. Egmond.; Gerard van Koten.; Jan-Erling Bäckvall.; Robertus J.M. Klein Gebbink. Covalent anchoring of a racemization catalyst to CALB-beads: towards dual immobilization of DKR catalysts.Tetrahedron Lett. 2011, 52 (14),1601-1604.
  2. Peter Blakskjæra.; Kurt V. Gothelf. Synthesis of an elongated linear oligo(phenylene ethynylene)-based building block for application in DNA-programmed assembly. Organic & Biomolecular Chemistry. 2006, 4 (18),3442-3447.