1-Boc-4-(3-hydroxypropyl)piperazine, 97%, Thermo Scientific Chemicals
1-Boc-4-(3-hydroxypropyl)piperazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-4-(3-hydroxypropyl)piperazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H55547.14
also known as H55547-14
Price (USD)/ Each
298.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
298.00
Add to cart
1-Boc-4-(3-hydroxypropyl)piperazine, 97%, Thermo Scientific Chemicals
Catalog numberH55547.14
Price (USD)/ Each
298.00
-
Add to cart
Chemical Identifiers
CAS132710-90-8
IUPAC Nametert-butyl 4-(3-hydroxypropyl)piperazine-1-carboxylate
Molecular FormulaC12H24N2O3
InChI KeyLRYRQGKGCIUVON-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCN(CCCO)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 96.0%
Appearance (Color)White to pale yellow
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only