2-Chloropyridine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
2-Chloropyridine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloropyridine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H55550.06
also known as H55550-06
Price (USD)/ Each
309.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
364.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
309.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
364.00 
Save 54.35 (15%)
Add to cart
2-Chloropyridine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
Catalog numberH55550.06
Price (USD)/ Each
309.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
364.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
CAS23100-12-1
IUPAC Name6-chloropyridine-3-carbaldehyde
Molecular FormulaC6H4ClNO
InChI KeyAFWWKZCPPRPDQK-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%
2-Chloropyridine-5-carboxaldehyde is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyridine-5-carboxaldehyde is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only