Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%
Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%
Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%
Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%
Thermo Scientific Chemicals

Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%

CAS: 92466-70-1 | C4H4F6O3P+ | 245.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H55562.06
also known as H55562-06
Price (USD)
89.60
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
89.60
Each
Add to cart
Specifications
CAS92466-70-1
Chemical Name or MaterialBis(2,2,2-trifluoroethyl) phosphite
Density1.545 g/mL
Flash Point76°C (169°F)
Formula Weight246.05
View more
Ask our AI about this Product
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Guy D. Joly.; Eric N. Jacobsen. Thiourea-Catalyzed Enantioselective Hydrophosphonylation of Imines: Practical Access to Enantiomerically Enriched α-Amino Phosphonic Acids. J. Am. Chem. Soc. 2004, 126 (13),4102-4103.
  2. Richard J Cohen.; Daniel L Fox.; Jarrod F Eubank.; Ralph Nicholas Salvatore. Mild and efficient Cs2CO3-promoted synthesis of phosphonates. Tetrahedron Lett. 2003, 44 (47),8617-8621.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link