Thermo Scientific Chemicals

Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals

Catalog number: H55596.03
1 g, Each
Thermo Scientific Chemicals

Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals

Catalog number: H55596.03
1 g, Each
Quantity
Catalog number: H55596.03
also known as H55596-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
960-16-7
IUPAC Name
(2,3,4-tributylphenyl)stannanylidyne
Molecular Formula
C18H29Sn
InChI Key
ZMUNITMSSGNSOT-UHFFFAOYSA-N
SMILES
CCCCC1=CC=C([Sn])C(CCCC)=C1CCCC
Appearance (Color)
Colorless
Assay (GC)
≥96.0%
Proton NMR
Conforms
Form
Liquid

Description

It is applied in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in chemical research.

Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text