4-Hydroxy-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
4-Hydroxy-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55597.03
also known as H55597-03
Price (USD)/ Each
37.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
37.20
Add to cart
4-Hydroxy-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberH55597.03
Price (USD)/ Each
37.20
-
Add to cart
Chemical Identifiers
CAS3272-08-0
IUPAC Name4-cyano-2-nitrobenzen-1-olate
Molecular FormulaC7H3N2O3
InChI KeyINBLGVOPOSGVTA-UHFFFAOYSA-M
SMILES[O-]C1=CC=C(C=C1[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only