2-Chloro-5-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
2-Chloro-5-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H55618.03
also known as H55618-03
Price (USD)/ Each
463.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
463.00
Add to cart
2-Chloro-5-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH55618.03
Price (USD)/ Each
463.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or Material2-Chloro-5-(methylcarbamoyl)benzeneboronic acid
CAS2096338-17-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Cécile Enguehard.; Jean-Louis Renou.; Valérie Collot.; Maud Hervet.; Sylvain Rault.; Alain Gueiffier. Reactivity of 3-Iodoimidazo[1,2-a]pyridines Using a Suzuki-Type Cross-Coupling Reaction.J. Org. Chem. 2000, 65 (20), 6572-6575.