Thermo Scientific Chemicals

Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals

Catalog number: H55634.03
1 g, Each
Thermo Scientific Chemicals

Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals

Catalog number: H55634.03
1 g, Each
Quantity
Catalog number: H55634.03
also known as H55634-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17846-68-3
IUPAC Name
azidotributylstannane
Molecular Formula
C12H29N3Sn
InChI Key
VJJINBNRYCPOOA-UHFFFAOYSA-N
SMILES
N=[N+]=[N-].CCCC[SnH](CCCC)CCCC
Infrared spectrum
Conforms
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay by NMR
≥94.0%

Description

Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

Solubility
Miscible with toluene, hexane, acetonitrile and tetrahydrofuran.

Notes
Use in a fume hood. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text