Thermo Scientific Chemicals

Triphenyl borate, 97%, Thermo Scientific Chemicals

Catalog number: H55686.06
5 g, Each
Thermo Scientific Chemicals

Triphenyl borate, 97%, Thermo Scientific Chemicals

Catalog number: H55686.06
5 g, Each
Quantity
Catalog number: H55686.06
also known as H55686-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1095-03-0
IUPAC Name
triphenyl borate
Molecular Formula
C18H15BO3
InChI Key
MDCWDBMBZLORER-UHFFFAOYSA-N
SMILES
O(B(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1
Purity
≥96.0%
Form
Powder
Appearance (Color)
Pale yellow to yellow

Description

Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

Solubility
Soluble in chloroform, methanol.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text