Triphenylantimony(V) dichloride, 99%, Thermo Scientific Chemicals
Triphenylantimony(V) dichloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triphenylantimony(V) dichloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H55719.14
also known as H55719-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS594-31-0
Molecular FormulaC18H17Cl2Sb
InChI KeyWSTBPJVOVGALEQ-UHFFFAOYSA-N
SMILESCl.Cl.C1=CC=C(C=C1)[Sb](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)425.99
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisPASS
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only