Thermo Scientific Chemicals

Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H55752.06
5 g, Each
Thermo Scientific Chemicals

Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H55752.06
5 g, Each
Quantity
Catalog number: H55752.06
also known as H55752-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1149-23-1
IUPAC Name
3,5-diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C13H19NO4
InChI Key
LJXTYJXBORAIHX-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=C(C)NC(C)=C(C1)C(=O)OCC
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥97.5% to ≤102.5% (Titration)
Appearance (Color)
Pale yellow to yellow to green

Description

Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.

Solubility
Soluble in organic solvents.

Notes
Desiccate at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text