(3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%, Thermo Scientific Chemicals
(3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H55754.14
also known as H55754-14
Price (USD)/ Each
593.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
593.00
Add to cart
(3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%, Thermo Scientific Chemicals
Catalog numberH55754.14
Price (USD)/ Each
593.00
-
Add to cart
Chemical Identifiers
CAS54314-85-1
IUPAC Name[3-(benzyloxy)propyl]triphenylphosphanium bromide
Molecular FormulaC28H28BrOP
InChI KeyDWYCJWXMZGVGJV-UHFFFAOYSA-M
SMILES[Br-].C(COCC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms
Appearance (Color)White
FormSolid
Purity≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only