4-Bromo-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific Chemicals
4-Bromo-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H55757.03
also known as H55757-03
Price (USD)/ Each
401.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
401.00
Add to cart
4-Bromo-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific Chemicals
Catalog numberH55757.03
Price (USD)/ Each
401.00
-
Add to cart
Chemical Identifiers
CAS175357-73-0
IUPAC Name4-bromo-N-[(4-methoxyphenyl)methyl]aniline
Molecular FormulaC14H14BrNO
InChI KeyLJKLYFDMLGYRCA-UHFFFAOYSA-N
SMILESCOC1=CC=C(CNC2=CC=C(Br)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only