Thermo Scientific Chemicals

2,6-Di-tert-butylpyridine, 97+%, Thermo Scientific Chemicals

Catalog number: H55761.03
1 g, Each
Thermo Scientific Chemicals

2,6-Di-tert-butylpyridine, 97+%, Thermo Scientific Chemicals

Catalog number: H55761.03
1 g, Each
Quantity
Catalog number: H55761.03
also known as H55761-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
585-48-8
IUPAC Name
2,6-di-tert-butylpyridine
Molecular Formula
C13H21N
InChI Key
UWKQJZCTQGMHKD-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=CC(=N1)C(C)(C)C
Appearance (Color)
Colorless to light yellow
Form
Liquid
Assay (HPLC)
>97.0%

Description

2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.

Solubility
Miscible with alcohol, acetone, and hexane. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text