Thermo Scientific Chemicals

9-Aminophenanthrene, 96%, Thermo Scientific Chemicals

Catalog number: H55773.03
1 g, Each
Thermo Scientific Chemicals

9-Aminophenanthrene, 96%, Thermo Scientific Chemicals

Catalog number: H55773.03
1 g, Each
Quantity
Catalog number: H55773.03
also known as H55773-03
Price (USD)
Quantity
-
Have Questions?

Description

9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

Solubility
It is slightly soluble in water 0.00581 mg/mL.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text